missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras-related protein Rab-11A Protein

Product Code. 16094210
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16094210

Brand: enQuireBio™ QP6566ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-11A, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001193765.1
Gene ID (Entrez) 8766
Name Ras-related protein Rab-11A Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol RAB11A
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence GTRDDEYDYL FKVVLIGDSG VGKSNLLSRF TRNEFNLESK STIGVEFATR SIQVDGKTIK AQIWDTAGQE RYRAITSAYY RGAVGALLVY DIAKHLTYEN VERWLKELRD HADSNIVIML VGNKSDLRHL RAVPTDEARA FAEKNGLSFI ETSALDSTNV EAAFQTILTE IYRIVSQKQM SDRRENDMSP SNNVVPIHVP PTTENKPKVQ CC
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.