missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-tyrosine sulfotransferase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-tyrosine sulfotransferase 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6834-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001008566.1 | |
Protein-tyrosine sulfotransferase 2 Protein | |
Research Use Only | |
TPST2 | |
Recombinant Protein | |
E. coli | |
QQVLECRAVL AGLRSPRGAM RPEQEELVMV GTNHVEYRYG KAMPLIFVGG VPRSGTTLMR AMLDAHPEVR CGEETRIIPR VLAMRQAWSK SGREKLRLDE AGVTDEVLDA AMQAFILEVI AKHGEPARVL CNKDPFTLKS SVYLSRLFPN SKFLLMVRDG RASVHSMITR KVTIAGFDLS SYRDCLTKWN KAIEVMYAQC MEVGKEKCLP VYYEQLVLHP RRSLKLILDF LGIAWSDAVL HHEDLIGKPG GVSLSKIERS TDQVIKPVNL EALSKWTGHI PGDVVRDMAQ IAPMLAQLGY DPYANPPNYG NPDPFVINNT QRVLKGDYKT PANLKGYFQV NQNSTSSHLG SS | |
Greater than 90% as determined by SDS-PAGE. |
8459 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |