missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-tyrosine sulfotransferase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-tyrosine sulfotransferase 2, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001008566.1 |
---|---|
Gene ID (Entrez) | 8459 |
Name | Protein-tyrosine sulfotransferase 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016160
|
enQuireBio™
QP6834-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16036160
|
enQuireBio™
QP6834-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16026160
|
enQuireBio™
QP6834-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16046160
|
enQuireBio™
QP6834-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16056160
|
enQuireBio™
QP6834-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16066160
|
enQuireBio™
QP6834-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001008566.1 | |
Protein-tyrosine sulfotransferase 2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
8459 | |
Research Use Only | |
TPST2 | |
Recombinant Protein | |
E. coli | |
QQVLECRAVL AGLRSPRGAM RPEQEELVMV GTNHVEYRYG KAMPLIFVGG VPRSGTTLMR AMLDAHPEVR CGEETRIIPR VLAMRQAWSK SGREKLRLDE AGVTDEVLDA AMQAFILEVI AKHGEPARVL CNKDPFTLKS SVYLSRLFPN SKFLLMVRDG RASVHSMITR KVTIAGFDLS SYRDCLTKWN KAIEVMYAQC MEVGKEKCLP VYYEQLVLHP RRSLKLILDF LGIAWSDAVL HHEDLIGKPG GVSLSKIERS TDQVIKPVNL EALSKWTGHI PGDVVRDMAQ IAPMLAQLGY DPYANPPNYG NPDPFVINNT QRVLKGDYKT PANLKGYFQV NQNSTSSHLG SS | |
Greater than 90% as determined by SDS-PAGE. |