missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP2CB Protein
A DNA sequence encoding the Homo sapiens (Human) PPP2CB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8342-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001009552.1 | |
PPP2CB Protein | |
Research Use Only | |
PPP2CB | |
Recombinant Protein | |
E. coli | |
MDDKAFTKEL DQWVEQLNEC KQLNENQVRT LCEKAKEILT KESNVQEVRC PVTVCGDVHG QFHDLMELFR IGGKSPDTNY LFMGDYVDRG YYSVETVTLL VALKVRYPER ITILRGNHES RQITQVYGFY DECLRKYGNA NVWKYFTDLF DYLPLTALVD GQIFCLHGGL SPSIDTLDHI RALDRLQEVP HEGPMCDLLW SDPDDRGGWG ISPRGAGYTF GQDISETFNH ANGLTLVSRA HQLVMEGYNW CHDRNVVTIF SAPNYCYRCG NQAAIMELDD TLKYSFLQFD PAPRRGEPHV TRRTPDYFL | |
Greater than 90% as determined by SDS-PAGE. |
5516 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |