missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP2CB Protein
A DNA sequence encoding the Homo sapiens (Human) PPP2CB, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001009552.1 |
---|---|
Gene ID (Entrez) | 5516 |
Name | PPP2CB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16014121
|
enQuireBio™
QP8342-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16034121
|
enQuireBio™
QP8342-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16024121
|
enQuireBio™
QP8342-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16044121
|
enQuireBio™
QP8342-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16054121
|
enQuireBio™
QP8342-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16064121
|
enQuireBio™
QP8342-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001009552.1 | |
PPP2CB Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5516 | |
Research Use Only | |
PPP2CB | |
Recombinant Protein | |
E. coli | |
MDDKAFTKEL DQWVEQLNEC KQLNENQVRT LCEKAKEILT KESNVQEVRC PVTVCGDVHG QFHDLMELFR IGGKSPDTNY LFMGDYVDRG YYSVETVTLL VALKVRYPER ITILRGNHES RQITQVYGFY DECLRKYGNA NVWKYFTDLF DYLPLTALVD GQIFCLHGGL SPSIDTLDHI RALDRLQEVP HEGPMCDLLW SDPDDRGGWG ISPRGAGYTF GQDISETFNH ANGLTLVSRA HQLVMEGYNW CHDRNVVTIF SAPNYCYRCG NQAAIMELDD TLKYSFLQFD PAPRRGEPHV TRRTPDYFL | |
Greater than 90% as determined by SDS-PAGE. |