missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPP2CB Protein

A DNA sequence encoding the Homo sapiens (Human) PPP2CB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8342-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024121

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001009552.1
PPP2CB Protein
Research Use Only
PPP2CB
Recombinant Protein
E. coli
MDDKAFTKEL DQWVEQLNEC KQLNENQVRT LCEKAKEILT KESNVQEVRC PVTVCGDVHG QFHDLMELFR IGGKSPDTNY LFMGDYVDRG YYSVETVTLL VALKVRYPER ITILRGNHES RQITQVYGFY DECLRKYGNA NVWKYFTDLF DYLPLTALVD GQIFCLHGGL SPSIDTLDHI RALDRLQEVP HEGPMCDLLW SDPDDRGGWG ISPRGAGYTF GQDISETFNH ANGLTLVSRA HQLVMEGYNW CHDRNVVTIF SAPNYCYRCG NQAAIMELDD TLKYSFLQFD PAPRRGEPHV TRRTPDYFL
Greater than 90% as determined by SDS-PAGE.
5516
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers