missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPARG Protein
A DNA sequence encoding the Homo sapiens (Human) PPARG, was expressed in the hosts and tags indicated.
287.00€ - 3175.00€
Specifications
Accession Number | NP_005028.4 |
---|---|
Gene ID (Entrez) | 5468 |
Name | PPARG Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023870
|
enQuireBio™
QP6518-YE-100UG |
100 μg |
920.00€
100µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16063870
|
enQuireBio™
QP6518-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16033870
|
enQuireBio™
QP6518-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16053870
|
enQuireBio™
QP6518-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16073870
|
enQuireBio™
QP6518-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16043870
|
enQuireBio™
QP6518-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_005028.4 | |
PPARG Protein | |
Not determined. | |
Full Length of Isoform 1 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5468 | |
Research Use Only | |
PPARG | |
Recombinant Protein | |
Yeast | |
MGETLGDSPI DPESDSFTDT LSANISQEMT MVDTEMPFWP TNFGISSVDL SVMEDHSHSF DIKPFTTVDF SSISTPHYED IPFTRTDPVV ADYKYDLKLQ EYQSAIKVEP ASPPYYSEKT QLYNKPHEEP SNSLMAIECR VCGDKASGFH YGVHACEGCK GFFRRTIRLK LIYDRCDLNC RIHKKSRNKC QYCRFQKCLA VGMSHNAIRF GRMPQAEKEK LLAEISSDID QLNPESADLR ALAKHLYDSY IKSFPLTKAK ARAILTGKTT DKSPFVIYDM NSLMMGEDKI KFKHITPLQE QSKEVAIRIF QGCQFRSVEA VQEITEYAKS IPGFVNLDLN DQVTLLKYGV HEIIYTMLAS LMNKDGVLIS EGQGFMTREF LKSLRKPFGD FMEPKFEFAV KFNALELDDS DLAIFIAVII LSGDRPGLLN VKPIEDIQDN LLQALELQLK LNHPESSQLF AKLLQKMTDL RQIVTEH | |
Greater than 80% as determined by SDS-PAGE. |