missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PLA2G7/PAF-AH/Lp-PLA2 Protein
Click to view available options
:
10ug; Carrier Free
Description
The Recombinant Human PLA2G7/PAF-AH/Lp-PLA2 Protein is derived from E. coli. The Recombinant Human PLA2G7/PAF-AH/Lp-PLA2 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Sodium Acetate, NaCl and Glycerol. |
| Gene ID (Entrez) | 7941 |
| Molecular Weight (g/mol) | Predicted MW: 46kDa Observed MW: 42 - 45kDa, reducing conditions |
| Name | PLA2G7/PAF-AH/Lp-PLA2 |
| Quantity | 10μg |
| Source | E. coli-derived human PLA2G7/PAF-AH/Lp-PLA2 protein Met33-Asn441 & Leu45-Asn441,both with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction