missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Peroxiredoxin 2 Protein

Product Code. p-7145933 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
This item is not returnable. View return policy

Product Code. 18230663

Brand: Novus Biologicals™ NBP261185100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-198 of Mouse Peroxiredoxin 2alpha The Recombinant Mouse Peroxiredoxin 2 Protein is derived from E. coli. The Recombinant Mouse Peroxiredoxin 2 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/ml
For Use With (Application) Bioactivity, Functional Study, SDS-PAGE
Formulation PBS (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 7001
Molecular Weight (g/mol) 24.3 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol PRDX2
Cross Reactivity Peroxiredoxin 2
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.