missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nucleoredoxin-like protein 1

Product Code. 16000351
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16000351 1 mg 1mg
16090341 10 μg 10µg
16030351 50 μg 50µg
16080341 100 μg 100µg
16010351 200 μg 200µg
16020351 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16000351 Supplier enQuireBio™ Supplier No. QP7712ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Nucleoredoxin-like protein 2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001155097.1
Gene ID (Entrez) 158046
Name Nucleoredoxin-like protein 1
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol NXNL2
Protein Length Full Length of Isoform 2
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MVDILGERHL VTCKGATVEA EAALQNKVVA LYFAAARCAP SRDFTPLLCD FYTALVAEAR RPAPFEVVFV SADGSCQEML DFMRELHGAW LALPFHDPYR QRSLALLPRL ECSGVILAHC NLCLLGSSDS LALAS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.