missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nucleolar protein 58 Protein

A DNA sequence encoding the Homo sapiens (Human) Nucleolar protein 58, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8278-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16063731

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_057018.1
Nucleolar protein 58 Protein
Research Use Only
NOP58
Recombinant Protein
E. coli
MLVLFETSVG YAIFKVLNEK KLQEVDSLWK EFETPEKANK IVKLKHFEKF QDTAEALAAF TALMEGKINK QLKKVLKKIV KEAHEPLAVA DAKLGGVIKE KLNLSCIHSP VVNELMRGIR SQMDGLIPGV EPREMAAMCL GLAHSLSRYR LKFSADKVDT MIVQAISLLD DLDKELNNYI MRCREWYGWH FPELGKIISD NLTYCKCLQK VGDRKNYASA KLSELLPEEV EAEVKAAAEI SMGTEVSEED ICNILHLCTQ VIEISEYRTQ LYEYLQNRMM AIAPNVTVMV GELVGARLIA HAGSLLNLAK HAASTVQILG AEKALFRALK SRRDTPKYGL IYHASLVGQT SPKHKGKISR MLAAKTVLAI RYDAFGEDSS SAMGVENRAK LEARLRTLED RGIRKISGTG KALAKTEKYE HKSEVKTYDP SGDSTLPTCS KKRKIEQVDK EDEITEKKAK KAKIKVKVEE EEEEKVAEEE ETSVKKKKKR GKKKHIKEEP LSEEEPCTST AIASPEKKKK KKKKRENED
Greater than 90% as determined by SDS-PAGE.
51602
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers