missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleolar protein 58 Protein
A DNA sequence encoding the Homo sapiens (Human) Nucleolar protein 58, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_057018.1 |
---|---|
Gene ID (Entrez) | 51602 |
Name | Nucleolar protein 58 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013731
|
enQuireBio™
QP8278-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
16033731
|
enQuireBio™
QP8278-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
16023731
|
enQuireBio™
QP8278-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16043731
|
enQuireBio™
QP8278-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16053731
|
enQuireBio™
QP8278-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16063731
|
enQuireBio™
QP8278-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_057018.1 | |
Nucleolar protein 58 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
51602 | |
Research Use Only | |
NOP58 | |
Recombinant Protein | |
E. coli | |
MLVLFETSVG YAIFKVLNEK KLQEVDSLWK EFETPEKANK IVKLKHFEKF QDTAEALAAF TALMEGKINK QLKKVLKKIV KEAHEPLAVA DAKLGGVIKE KLNLSCIHSP VVNELMRGIR SQMDGLIPGV EPREMAAMCL GLAHSLSRYR LKFSADKVDT MIVQAISLLD DLDKELNNYI MRCREWYGWH FPELGKIISD NLTYCKCLQK VGDRKNYASA KLSELLPEEV EAEVKAAAEI SMGTEVSEED ICNILHLCTQ VIEISEYRTQ LYEYLQNRMM AIAPNVTVMV GELVGARLIA HAGSLLNLAK HAASTVQILG AEKALFRALK SRRDTPKYGL IYHASLVGQT SPKHKGKISR MLAAKTVLAI RYDAFGEDSS SAMGVENRAK LEARLRTLED RGIRKISGTG KALAKTEKYE HKSEVKTYDP SGDSTLPTCS KKRKIEQVDK EDEITEKKAK KAKIKVKVEE EEEEKVAEEE ETSVKKKKKR GKKKHIKEEP LSEEEPCTST AIASPEKKKK KKKKRENED | |
Greater than 90% as determined by SDS-PAGE. |