Learn More
Abcam Recombinant Human NT-ProANP Protein Standard
Product Code. 32141515
Change view
Click to view available options
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
32141515 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32141515 Supplier Abcam Supplier No. ab31669610ug
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Recombinant Human NT-ProANP Protein Standard is a Human Fragment protein, expressed in HEK 293 cells, with >80%, suitable for sELISA, SDS-PAGE.
Recombinant Human NT-ProANP Protein Standard is a Human Fragment protein, expressed in HEK 293 cells, with >80%, suitable for sELISA, SDS-PAGE.
The product is subject to the following: Abcam Restricted Use Statement