Learn More
Abcam Recombinant Human NT-ProANP Protein Standard
Description
Recombinant Human NT-ProANP Protein Standard is a Human Fragment protein, expressed in HEK 293 cells, with >80%, suitable for sELISA, SDS-PAGE.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 10 μg |
| Regulatory Status | RUO |