missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NET1 Protein

A DNA sequence encoding the Homo sapiens (Human) NET1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7782-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020771

  • 2385.00€ / 1mg
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001040625.1
NET1 Protein
Research Use Only
NET1
Recombinant Protein
E. coli
MVAHDETGGL LPIKRTIRVL DVNNQSFREQ EEPSNKRVRP LARVTSLANL ISPVRNGAVR RFGQTIQSFT LRGDHRSPAS AQKFSSRSTV PTPAKRRSSA LWSEMLDITM KESLTTREIR RQEAIYEMSR GEQDLIEDLK LARKAYHDPM LKLSIMSEEE LTHIFGDLDS YIPLHEDLLT RIGEATKPDG TVEQIGHILV SWLPRLNAYR GYCSNQLAAK ALLDQKKQDP RVQDFLQRCL ESPFSRKLDL WSFLDIPRSR LVKYPLLLKE ILKHTPKEHP DVQLLEDAIL IIQGVLSDIN LKKGESECQY YIDKLEYLDE KQRDPRIEAS KVLLCHGELR SKSGHKLYIF LFQDILVLTR PVTRNERHSY QVYRQPIPVQ ELVLEDLQDG DVRMGGSFRG AFSNSEKAKN IFRIRFHDPS PAQSHTLQAN DVFHKQQWFN CIRAAIAPFQ SAGSPPELQG LPELHEECEG NHPSARKLTA QRRASTVSSV TQVEVDENAY RCGSGMQMAE DSKSLKTHQT QPGIRRARDK ALSGGKRKET LV
Greater than 90% as determined by SDS-PAGE.
10276
1 mg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers