missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NET1 Protein
A DNA sequence encoding the Homo sapiens (Human) NET1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001040625.1 |
---|---|
Gene ID (Entrez) | 10276 |
Name | NET1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16000771
|
enQuireBio™
QP7782-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16020771
|
enQuireBio™
QP7782-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16010771
|
enQuireBio™
QP7782-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16030771
|
enQuireBio™
QP7782-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16040771
|
enQuireBio™
QP7782-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16050771
|
enQuireBio™
QP7782-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001040625.1 | |
NET1 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10276 | |
Research Use Only | |
NET1 | |
Recombinant Protein | |
E. coli | |
MVAHDETGGL LPIKRTIRVL DVNNQSFREQ EEPSNKRVRP LARVTSLANL ISPVRNGAVR RFGQTIQSFT LRGDHRSPAS AQKFSSRSTV PTPAKRRSSA LWSEMLDITM KESLTTREIR RQEAIYEMSR GEQDLIEDLK LARKAYHDPM LKLSIMSEEE LTHIFGDLDS YIPLHEDLLT RIGEATKPDG TVEQIGHILV SWLPRLNAYR GYCSNQLAAK ALLDQKKQDP RVQDFLQRCL ESPFSRKLDL WSFLDIPRSR LVKYPLLLKE ILKHTPKEHP DVQLLEDAIL IIQGVLSDIN LKKGESECQY YIDKLEYLDE KQRDPRIEAS KVLLCHGELR SKSGHKLYIF LFQDILVLTR PVTRNERHSY QVYRQPIPVQ ELVLEDLQDG DVRMGGSFRG AFSNSEKAKN IFRIRFHDPS PAQSHTLQAN DVFHKQQWFN CIRAAIAPFQ SAGSPPELQG LPELHEECEG NHPSARKLTA QRRASTVSSV TQVEVDENAY RCGSGMQMAE DSKSLKTHQT QPGIRRARDK ALSGGKRKET LV | |
Greater than 90% as determined by SDS-PAGE. |