missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NET1 Protein

A DNA sequence encoding the Homo sapiens (Human) NET1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7782-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040771

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001040625.1
NET1 Protein
Research Use Only
NET1
Recombinant Protein
E. coli
MVAHDETGGL LPIKRTIRVL DVNNQSFREQ EEPSNKRVRP LARVTSLANL ISPVRNGAVR RFGQTIQSFT LRGDHRSPAS AQKFSSRSTV PTPAKRRSSA LWSEMLDITM KESLTTREIR RQEAIYEMSR GEQDLIEDLK LARKAYHDPM LKLSIMSEEE LTHIFGDLDS YIPLHEDLLT RIGEATKPDG TVEQIGHILV SWLPRLNAYR GYCSNQLAAK ALLDQKKQDP RVQDFLQRCL ESPFSRKLDL WSFLDIPRSR LVKYPLLLKE ILKHTPKEHP DVQLLEDAIL IIQGVLSDIN LKKGESECQY YIDKLEYLDE KQRDPRIEAS KVLLCHGELR SKSGHKLYIF LFQDILVLTR PVTRNERHSY QVYRQPIPVQ ELVLEDLQDG DVRMGGSFRG AFSNSEKAKN IFRIRFHDPS PAQSHTLQAN DVFHKQQWFN CIRAAIAPFQ SAGSPPELQG LPELHEECEG NHPSARKLTA QRRASTVSSV TQVEVDENAY RCGSGMQMAE DSKSLKTHQT QPGIRRARDK ALSGGKRKET LV
Greater than 90% as determined by SDS-PAGE.
10276
500 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers