missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NET1 Protein
A DNA sequence encoding the Homo sapiens (Human) NET1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7782-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001040625.1 | |
NET1 Protein | |
Research Use Only | |
NET1 | |
Recombinant Protein | |
E. coli | |
MVAHDETGGL LPIKRTIRVL DVNNQSFREQ EEPSNKRVRP LARVTSLANL ISPVRNGAVR RFGQTIQSFT LRGDHRSPAS AQKFSSRSTV PTPAKRRSSA LWSEMLDITM KESLTTREIR RQEAIYEMSR GEQDLIEDLK LARKAYHDPM LKLSIMSEEE LTHIFGDLDS YIPLHEDLLT RIGEATKPDG TVEQIGHILV SWLPRLNAYR GYCSNQLAAK ALLDQKKQDP RVQDFLQRCL ESPFSRKLDL WSFLDIPRSR LVKYPLLLKE ILKHTPKEHP DVQLLEDAIL IIQGVLSDIN LKKGESECQY YIDKLEYLDE KQRDPRIEAS KVLLCHGELR SKSGHKLYIF LFQDILVLTR PVTRNERHSY QVYRQPIPVQ ELVLEDLQDG DVRMGGSFRG AFSNSEKAKN IFRIRFHDPS PAQSHTLQAN DVFHKQQWFN CIRAAIAPFQ SAGSPPELQG LPELHEECEG NHPSARKLTA QRRASTVSSV TQVEVDENAY RCGSGMQMAE DSKSLKTHQT QPGIRRARDK ALSGGKRKET LV | |
Greater than 90% as determined by SDS-PAGE. |
10276 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |