missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Nectin-1 Fc Chimera Protein, CF
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human Nectin-3 (Catalog # 3064-N3 ) is immobilized at 1 μg/mL (100 μL/well), Recombinant Human Nectin-1 Fc Chimera (Catalog # 10697-N1) binds with an ED50 of 2.5-20 ng/mL.
Specifications
Specifications
| Accession Number | Q15223.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 5818 |
| Molecular Weight (g/mol) | M.W. (Observed): 78-88 kDa, under reducing conditions, M.W. (Theoretical): 83 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Nectin-1 protein Human Nectin-1 (Gln31-Gly346) (N-terminus) Accession # Q15223.3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin1, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1, PRR1MGC16207, PVRL1, PVRR1, PVRR1nectin-1, SK-12 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction