missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Myoglobin Protein

Product Code. 18639407 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18639407 1 mg 1mg
18686087 500 μg 500µg
18642467 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18639407 Supplier Novus Biologicals™ Supplier No. NBP3071081mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifikationer

Formulation 0.2 μm filtered solution in 25 mM Tris-HCl (pH 8.0)
Gene ID (Entrez) 4151
Molecular Weight (g/mol) MolecularWeight-theroretical: 17.1 kDa
Quantity 1 mg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1.0 EU/μg of Myoglobin as determined by LAL method.
Gene Alias MGC13548, myoglobin, PVALB
Gene Symbol MB
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Form Purified
Research Category Cancer, Cardiovascular Biology
Purity or Quality Grade >95%, by SDS-PAGE
Protein Myoglobin
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.