missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Myoglobin Protein

Product Code. 18686087 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18686087 500 μg 500µg
18639407 1 mg 1mg
18642467 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18686087 Supplier Novus Biologicals™ Supplier No. NBP307108500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Formulation 0.2 μm filtered solution in 25 mM Tris-HCl (pH 8.0)
Gene ID (Entrez) 4151
Molecular Weight (g/mol) MolecularWeight-theroretical: 17.1 kDa
Quantity 500 μg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1.0 EU/μg of Myoglobin as determined by LAL method.
Gene Alias MGC13548, myoglobin, PVALB
Gene Symbol MB
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Form Purified
Research Category Cancer, Cardiovascular Biology
Purity or Quality Grade >95%, by SDS-PAGE
Protein Myoglobin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.