missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human MTMR1 GST (N-Term) Protein

Product Code. 18304019 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
18304019 10 μg 10µg
18328049 25 μg 25µg
2 options
This item is not returnable. View return policy

Product Code. 18304019

Brand: Novus Biologicals™ H00008776P0110ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 8776
Molecular Weight (g/mol) 29.92 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias EC 3.1.3, EC 3.1.3.-, EC 3.1.3.48, myotubularin related protein 1, myotubularin-related protein 1
Gene Symbol MTMR1
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein MTMR1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.