missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat GDF-8/Myostatin Protein, CF

Product Code. 30306772
Click to view available options
Quantity:
100 μg
20 μg
250 μg
50 μg
Unit Size:
100µg
20µg
250µg
50µg
This item is not returnable. View return policy

Product Code. 30306772

Brand: R&D Systems™ 11658G8100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to induce cytotoxicity using MPC-11 mouse B lymphocyte cells. The ED50 for this effect is 4.00-40.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14793.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 2660
Molecular Weight (g/mol) MolecularWeight-observed: 10-14 kDa, under reducing conditions., MolecularWeight-theroretical: 12 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived GDF-8/Myostatin protein Asp267-Ser375
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water.
Protein GDF-8/Myostatin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.