missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat GDF-8/Myostatin Protein

Product Code. 18717006
Click to view available options
Quantity:
100 μg
250 μg
Unit Size:
100µg
250µg
This item is not returnable. View return policy

Product Code. 18717006

Brand: R&D Systems™ 788G8100/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. The ED50 for this effect is 2 to 10 ng/mL.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number O08689
For Use With (Application) Bioactivity
Gene ID (Entrez) 2660
Molecular Weight (g/mol) 12.4 kDa
Name GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived GDF-8/Myostatin protein Asp268-Ser376
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol MSTN
Conjugate Unconjugated
Species Human, Mouse, Rat
Reconstitution Reconstitute at 100 μg/mL in sterile 4 mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in Acetonitrile and TFA.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.