missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human MBL His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1mg
0.5 mg
Unit Size:
0.1mg
0.5mg
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 108-248 of Human MBL The Recombinant Human MBL Protein is derived from E. coli. The Recombinant Human MBL Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_000233 |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20 mM Tris-HCl buffer (pH 8.0), 2 M UREA, 20% glycerol, 0.2 M Nacl |
| Gene ID (Entrez) | 4153 |
| Molecular Weight (g/mol) | 18 kDa |
| Name | MBL Protein |
| Purification Method | SDS-PAGE |
| Quantity | 0.5 mg |
| Source | E. Coli |
| Immunogen | MBL2, 108-248 aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSAASERKA LQTEMARIKK WLTFSLGKQV GNKFFLTNGE IMTFEKVKAL CVKFQASVAT PRNAAENGAI QNLIKEEAFL GITDEKTEGQ FVDLTGNRLT YTNWNEGEPN NAGSDEDCVL LLKNGQWNDV PCSTSHLAVC EFPI |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction