Learn More
R&D Systems™ Recombinant Human LRP-1 Cluster IV Fc Chimera Protein
Click to view available options
:
50ug; Carrier Free
Beschreibung
Measured by its binding ability in a functional ELISA. In a Human IgG Fc Antibody (Catalog # G-102-C) coated plate, Recombinant Human LRP-1 Cluster IV Fc Chimera binds Recombinant Human LRPAP (Catalog # 4296-LR ) with an ED50 of 0.25-1.25 ng/mL.
Spezifikation
Spezifikation
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 4035 |
| Molecular Weight (g/mol) | 76.7 kDa |
| Name | LRP-1 Cluster IV Fc Chimera |
| Quantity | 50μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human LRP-1 Cluster IV protein, (N-terminus) Human LRP1-C4 (Ser3332-Asp3779) Accession &Num; Q07954 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur