missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human LDLR Protein

Product Code. 18338387
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18338387

Brand: R&D Systems™ 2148LD025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Human LDLR Protein is derived from NS0. The Recombinant Human LDLR Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 3949
Molecular Weight (g/mol) 85.6 kDa & 66.8 kDa
Name LDLR
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived human LDL R protein Ala22-Arg788,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.1 EU per 1μg of the protein by the LAL method.
Gene Alias FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.