missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLRG1 Protein
A DNA sequence encoding the Homo sapiens (Human) KLRG1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7938-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001316028.1 | |
KLRG1 Protein | |
Research Use Only | |
KLRG1 | |
Recombinant Protein | |
E. coli | |
LCQGSNYSTC ASCPSCPDRW MKYGNHCYYF SVEEKDWNSS LEFCLARDSH LLVITDNQEM SLLQVFLSEA FCWIGLRNNS GWRWEDGSPL NFSRISSNSF VQTCGAINKN GLQASSCEVP LHWVCKKCPF ADQALF | |
Greater than 90% as determined by SDS-PAGE. |
10219 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |