missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLRG1 Protein
A DNA sequence encoding the Homo sapiens (Human) KLRG1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001316028.1 |
---|---|
Gene ID (Entrez) | 10219 |
Name | KLRG1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011721
|
enQuireBio™
QP7938-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16031721
|
enQuireBio™
QP7938-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16021721
|
enQuireBio™
QP7938-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16041721
|
enQuireBio™
QP7938-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16051721
|
enQuireBio™
QP7938-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16061721
|
enQuireBio™
QP7938-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001316028.1 | |
KLRG1 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10219 | |
Research Use Only | |
KLRG1 | |
Recombinant Protein | |
E. coli | |
LCQGSNYSTC ASCPSCPDRW MKYGNHCYYF SVEEKDWNSS LEFCLARDSH LLVITDNQEM SLLQVFLSEA FCWIGLRNNS GWRWEDGSPL NFSRISSNSF VQTCGAINKN GLQASSCEVP LHWVCKKCPF ADQALF | |
Greater than 90% as determined by SDS-PAGE. |