missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KLRG1 Protein

A DNA sequence encoding the Homo sapiens (Human) KLRG1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7938-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041721

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001316028.1
KLRG1 Protein
Research Use Only
KLRG1
Recombinant Protein
E. coli
LCQGSNYSTC ASCPSCPDRW MKYGNHCYYF SVEEKDWNSS LEFCLARDSH LLVITDNQEM SLLQVFLSEA FCWIGLRNNS GWRWEDGSPL NFSRISSNSF VQTCGAINKN GLQASSCEVP LHWVCKKCPF ADQALF
Greater than 90% as determined by SDS-PAGE.
10219
200 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers