missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL2 / Interleukin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) IL2 / Interleukin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6216-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000577.2 | |
IL2 / Interleukin-2 Protein | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
E. coli | |
APTSSSTKKT QLQLEHLLLD LQMILNGINN YKNPKLTRML TFKFYMPKKA TELKHLQCLE EELKPLEEVL NLAQSKNFHL RPRDLISNIN VIVLELKGSE TTFMCEYADE TATIVEFLNR WITFCQSIIS TLT | |
Greater than 90% as determined by SDS-PAGE. |
3558 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |