missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL2 / Interleukin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) IL2 / Interleukin-2, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_000577.2 |
---|---|
Gene ID (Entrez) | 3558 |
Name | IL2 / Interleukin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031650
|
enQuireBio™
QP6216-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16051650
|
enQuireBio™
QP6216-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16041650
|
enQuireBio™
QP6216-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16061650
|
enQuireBio™
QP6216-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16071650
|
enQuireBio™
QP6216-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16081650
|
enQuireBio™
QP6216-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_000577.2 | |
IL2 / Interleukin-2 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3558 | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
E. coli | |
APTSSSTKKT QLQLEHLLLD LQMILNGINN YKNPKLTRML TFKFYMPKKA TELKHLQCLE EELKPLEEVL NLAQSKNFHL RPRDLISNIN VIVLELKGSE TTFMCEYADE TATIVEFLNR WITFCQSIIS TLT | |
Greater than 90% as determined by SDS-PAGE. |