missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-9 Protein
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
The Recombinant Human IL-9 Protein is derived from Sf 21 (stably transfected). The Recombinant Human IL-9 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | P15248 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS |
| Gene ID (Entrez) | 3578 |
| Molecular Weight (g/mol) | 12 to 24 kDa |
| Quantity | 10 μg |
| Source | Spodoptera frugiperda,Sf 21 (stably transfected)-derived human IL-9 protein Gln19-Ile144 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction