missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-9 Protein

Product Code. 18706227
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18706227

Brand: R&D Systems™ 209ILB010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-9 Protein is derived from Sf 21 (stably transfected). The Recombinant Human IL-9 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15248
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 3578
Molecular Weight (g/mol) 12 to 24 kDa
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (stably transfected)-derived human IL-9 protein Gln19-Ile144
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name Cytokine P40; HP40; IL9; IL-9; IL-9homolog of mouse T cell and mast cell growth factor 40; interleukin 9; interleukin-9; p40 cytokine; p40 T-cell and mast cell growth factor; P40; T-cell growth factor P40
Biological Activity 0.03 to 0.3ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.