missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-4R alpha Fc Biotinylated Protein, CF

Product Code. 30713258
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30713258 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30713258 Supplier R&D Systems™ Supplier No. BT7700050
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fc Chimera

Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human IL-4R alpha Fc Chimera binds to Recombinant Human IL-4 Protein (Catalog # 204-IL ) with an ED50 of 0.100-1.00 Âμg/mL.
TRUSTED_SUSTAINABILITY

Spécification

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3566
Molecular Weight (g/mol) M.W.-observed: 55-85 kDa, under reducing conditions., M.W.-theroretical: 51 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human IL-4R alpha protein Human IL-4R alpha (Gly24-His 232) (N-terminus)Accession # P24394.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
Conjugate Biotin
Reconstitution Reconstitute at 250 μg/mL in water.
Protein IL-4R alpha
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.