missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-11 Protein
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in PBS and EDTA.
Unit Size:
25µg
5µg
Description
Measured in a cell proliferation assay using T11 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.02-0.12 ng/mL.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in PBS and EDTA. |
| Gene ID (Entrez) | 3589 |
| Molecular Weight (g/mol) | 19 kDa |
| Name | IL-11 |
| Quantity | 5 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-11 protein Pro22-Leu199 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, 2 to 8 °C as supplied. |
| Gene Alias | Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction