missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-1 beta/IL-1F2 Protein

Product Code. 18317457
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. 1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS. See Certificate of Analysis for details.
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Unit Size:
25µg
5µg
This item is not returnable. View return policy

Product Code. 18317457

Brand: R&D Systems™ 201LB025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells. Symons, J.A. et al. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 272. The ED50 for this effect is <12 pg/mL. The specific activity of recombinant human IL-1 beta is approximately 1.4 x 105 IU/μg, which is calibrated against recombinant human IL-1 beta WHO International Standard (NIBSC code: 86/680). Specific activity is for reference purposes only and is not routinely tested.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. 1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS. See Certificate of Analysis for details.
Gene ID (Entrez) 3553
Molecular Weight (g/mol) 17 kDa
Name IL-1 beta/IL-1F2
Quantity 25 μg
Source E. coli-derived human IL-1 beta/IL-1F2 protein Ala117-Ser269
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. The 1 mg pack size (01M) can be stored for 1 month at 2 to 8 °C under sterile conditions after opening.
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 50-200 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.