missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human IGSF8/CD316 His Protein

Product Code. 18602517 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18602517

Brand: Novus Biologicals™ NBP25988850ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 30% glycerol
Gene ID (Entrez) 93185
Molecular Weight (g/mol) MolecularWeight-theroretical: 59.6 kDa
Quantity 50 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less Than 1.0 EU / 1 μg of protein (determined by LAL method)
Gene Alias CD316, CD316 antigen, CD81P3LIR-D1, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRLimmunoglobulin superfamily member 8
Gene Symbol IGSF8
Research Category Neuroscience
Purity or Quality Grade >95%, by SDS-PAGE
Protein IGSF8/CD316
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.