missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IGF-I R/IGF1R Fc Chimera Protein, CF
Click to view available options
:
50 μg
Description
Measured by its binding ability in a functional ELISA. Recombinant Human IGF-I R/IGF1R Fc Chimera (Catalog # 11655-GR binds to Recombinant Human IGF-I/IGF-1 (Catalog # 291-G1 ) with an ED50 of 3.00-45.0 ng/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3480 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 160-180 kDa, 110-125 kDa and 70-80 kDa, under reducing conditions, MolecularWeight-theroretical: 130 kDa (single chain), 80 kDa (alpha subunit) and 49 kDa (beta subunit) |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human IGF-I R/IGF1R protein Human IGF1R (Glu31-His935) (N-terminus) Accession # P08069.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction