missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human HVEM/TNFRSF14 hIgG-His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.01 mg
0.05 mg
Unit Size:
0.01mg
0.05mg
Description
A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 39-202 of Human HVEM/TNFRSF14 The Recombinant Human HVEM/TNFRSF14 Protein is derived from Baculovirus. The Recombinant Human HVEM/TNFRSF14 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.25mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Phosphate buffered saline (pH 7.4) containing 10% glycerol |
| Gene ID (Entrez) | 8764 |
| Molecular Weight (g/mol) | TMW: 44.6kDa |
| Quantity | 0.05 mg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 |
| Common Name | HVEM/TNFRSF14 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction