missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HVEM/TNFRSF14 Fc Alexa Fluor™ 647 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by flow cytometry for its ability to bind anti-humanHVEM/TNFRSF14 Antibody conjugated beads. The concentration of Recombinant Human HVEM/TNFRSF14 Fc Chimera Alexa Fluor™ 647 (Catalog # AFR11177) that produces 50% of the binding response is 1.00-10.0 ng/mL.
Specifications
Specifications
| Accession Number | Q92956.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 8764 |
| Molecular Weight (g/mol) | M.W.-observed: 55-75 kDa, under reducing conditions., M.W.-theroretical: 44 kDa |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human HVEM/TNFRSF14 protein Human HVEM/TNFRSF14 (Pro37-Val202) (C-terminus) Accession # Q92956.3 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction