missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human HMG-CoA Reductase/HMGCR Protein

Product Code. 18797106
Click to view available options
:
Quantity: 20μg
This item is not returnable. View return policy

Product Code. 18797106

Brand: R&D Systems™ 9264HM020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human HMG-CoA Reductase/HMGCR Protein is derived from E. coli. The Recombinant Human HMG-CoA Reductase/HMGCR Protein has been validated for the following applications: Enzyme Activity.

Order Info

The product is shipped at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04035
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2μm filtered solution in Tris, NaCl, DTT and Glycerol
Gene ID (Entrez) 3156
Molecular Weight (g/mol) 56 to 63 kDa, reducing conditions
Name 3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, HMGCR, Hydroxymethylglutaryl-CoA Reductase, LDLCQ3
Quantity 20μg
Source E. coli-derived human HMG-CoA Reductase/HMGCR protein Ser426-Ala888,with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Biological Activity 50 to 750ng/mL (ED 50)
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag No
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.