missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 3126 |
| Molecular Weight (g/mol) | 51.81 kDa |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | class II histocompatibility antigen HLA DR alpha, beta1-0307, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR4, DR-4, DRB1 transplantation antigen, DRB4, HLA class II histocompatibility antigen, DR beta 4 chain, HLA class II histocompatibility antigen, DRB1-12 beta chain, HLA class II histocompatibility antigen, DRB1-4 beta chain, HLA-DR4B, human leucocyte antigen DRB4, leucocyte antigen DRB1, leukocyte antigen, major histocompatibility complex, class II, DR beta 1, major histocompatibility complex, class II, DR beta 4, MHC class I antigen DRB1*12, MHC class I antigen DRB1*4, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*4, MHC class II antigen DRB4, MHC class II antigen HLA-DRB1, MHC class II antigen HLA-DR-beta, MHC class II HLA-DR-beta-7, MHC class2 antigen, MHC HLA DR-beta chain |
| Gene Symbol | HLA-DRB4 |
| Research Category | Asthma, Immunology |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?