missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone-lysine N-methyltransferase SETDB1 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone-lysine N-methyltransferase SETDB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7573-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001138887.1 | |
Histone-lysine N-methyltransferase SETDB1 Protein | |
Research Use Only | |
SETDB1 | |
Recombinant Protein | |
E. coli | |
MSSLPGCIGL DAATATVESE EIAELQQAVV EELGISMEEL RHFIDEELEK MDCVQQRKKQ LAELETWVIQ KESEVAHVDQ LFDDASRAVT NCESLVKDFY SKLGLQYRDS SSEDESSRPT EIIEIPDEDD DVLSIDSGDA GSRTPKDQKL REAMAALRKS AQDVQKFMDA VNKKSSSQDL HKGTLSQMSG ELSKDGDLIV SMRILGKKRT KTWHKGTLIA IQTVGPGKKY KVKFDNKGKS LLSGNHIAYD YHPPADKLYV GSRVVAKYKD GNQVWLYAGI VAETPNVKNK LRFLIFFDDG YASYVTQSEL YPICRPLKKT WEDIEDISCR DFIEEYVTAY PNRPMVLLKS GQLIKTEWEG TWWKSRVEEV DGSLVRILFL VLFFSTILEA EVGGGGT | |
Greater than 80% as determined by SDS-PAGE. |
9869 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |