missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone-lysine N-methyltransferase SETDB1 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone-lysine N-methyltransferase SETDB1, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_001138887.1 |
---|---|
Gene ID (Entrez) | 9869 |
Name | Histone-lysine N-methyltransferase SETDB1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009490
|
enQuireBio™
QP7573-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16029490
|
enQuireBio™
QP7573-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16019490
|
enQuireBio™
QP7573-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16039490
|
enQuireBio™
QP7573-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16049490
|
enQuireBio™
QP7573-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16059490
|
enQuireBio™
QP7573-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001138887.1 | |
Histone-lysine N-methyltransferase SETDB1 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9869 | |
Research Use Only | |
SETDB1 | |
Recombinant Protein | |
E. coli | |
MSSLPGCIGL DAATATVESE EIAELQQAVV EELGISMEEL RHFIDEELEK MDCVQQRKKQ LAELETWVIQ KESEVAHVDQ LFDDASRAVT NCESLVKDFY SKLGLQYRDS SSEDESSRPT EIIEIPDEDD DVLSIDSGDA GSRTPKDQKL REAMAALRKS AQDVQKFMDA VNKKSSSQDL HKGTLSQMSG ELSKDGDLIV SMRILGKKRT KTWHKGTLIA IQTVGPGKKY KVKFDNKGKS LLSGNHIAYD YHPPADKLYV GSRVVAKYKD GNQVWLYAGI VAETPNVKNK LRFLIFFDDG YASYVTQSEL YPICRPLKKT WEDIEDISCR DFIEEYVTAY PNRPMVLLKS GQLIKTEWEG TWWKSRVEEV DGSLVRILFL VLFFSTILEA EVGGGGT | |
Greater than 80% as determined by SDS-PAGE. |