Learn More
R&D Systems™ Recombinant Human Histone Deacetylase 8 Protein
Product Code. 18187040
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. 18187040
Brand: R&D Systems™ 4359DA050
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
The Recombinant Human Histone Deacetylase 8 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Histone Deacetylase 8 Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Supplied as a 0.2μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 55869
Molecular Weight (g/mol) 43 kDa
Name Histone Deacetylase 8/HDAC8
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Histone Deacetylase 8/HDAC8 protein Met1-Val377,with an N-terminal Met and 7-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.