missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HDAC4 Protein

A DNA sequence encoding the Homo sapiens (Human) HDAC4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8906-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097111

  • 458.00€ / 100µg

Estimated Shipment: 22-05-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_006028.2
HDAC4 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
9759
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HDAC4
Recombinant Protein
E. coli
MSSQSHPDGL SGRDQPVELL NPARVNHMPS TVDVATALPL QVAPSAVPMD LRLDHQFSLP VAEPALREQQ LQQELLALKQ KQQIQRQILI AEFQRQHEQL SRQHEAQLHE HIKQQQEMLA MKHQQELLEH QRKLERHRQE QELEKQHREQ KLQQLKNKEK GKESAVASTE VKMKLQEFVL NKKKALAHRN LNHCISSDPR YWYGKTQHSS LDQSSPPQSG VSTSYNH
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers