Learn More
R&D Systems™ Recombinant Human HAI-1 Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Recombinant Human HAI-1 Protein is derived from NS0. The Recombinant Human HAI-1 Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
| For Use With (Application) | Inhibition Assay |
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 6692 |
| Molecular Weight (g/mol) | 47 kDa |
| Name | HAI-1 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human HAI-1 protein Pro37-Glu449,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |