missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human GRP78/HSPA5 His Protein

Product Code. 18544624 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
This item is not returnable. View return policy

Product Code. 18544624

Brand: Novus Biologicals™ NBC1183780.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 20-650 of Human GRP78/HSPA5 The Recombinant Human GRP78/HSPA5 Protein is derived from E. coli. The Recombinant Human GRP78/HSPA5 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11021
Concentration 1 mg/mL
For Use With (Application) Flow Cytometry
Formulation 20 mM Tris-HCl buffer (pH 8.0), 10% glycerol
Gene ID (Entrez) 3309
Molecular Weight (g/mol) M.W. (theoretical): 71 kDa
Name GRP78/HSPA5 Protein
Purification Method Protein
Quantity 0.5 mg
Source E. Coli
Immunogen 20-650aa, Human, His-Tagged, Recombinant, E.coli (NP_005338.1).
Storage Requirements Store at 4°C short term. Aliquot and store (at -20°C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol HSPA5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Cancer, Unfolded Protein Response
Purity or Quality Grade >90%, by SDS-PAGE
Protein GRP78/HSPA5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.