missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GDNF Protein

Product Code. 18798229
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18798229

Brand: R&D Systems™ 212GD01M/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 2-12 ng/mL in the presence of Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ). Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ) at 1 μg/mL can bind Recombinant Human GDNF with an apparent Kd <1 nM.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P39905
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 2668
Molecular Weight (g/mol) 11.6 kDa (monomer)
Quantity 1 mg
Source Mouse myeloma cell line,NS0-derived human GDNF protein Arg109-Ile211
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method
Biological Activity 2 to 12ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.