missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GCC1 Protein
A DNA sequence encoding the Homo sapiens (Human) GCC1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7935-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_078799.2 | |
GCC1 Protein | |
Research Use Only | |
GCC1 | |
Recombinant Protein | |
E. coli | |
FGMNFGGGPS KKDLLETIET QKKQLLQYQA RLKDVVRAYK SLLKEKEALE ASIKVLSVSH EADVGLAGVQ LPGLTFPDSV DDRCSTHSED STGTATSLDT AASLTSTKGE FGVEDDRPAR GPPPPKSEEA SWSESGVSSS SGDGPFAGGE VDKRLHQLKT QLATLTSSLA TVTQEKSRME ASYLADKKKM KQDLEDASNK AEEERARLEG ELKGLQEQIA ETKARLITQQ HDRAQEQSDH ALMLRELQKL LQEERTQRQD LELRLEETRE ALAGRAYAAE QMEGFELQTK QLTREVEELK SELQAIRDEK NQPDPRLQEL QEEAARLKSH FQAQLQQEMR KTALAEDQLR QQSQVEEQRV AALENQISEV SELLGTYEKA KQKDQLAIQK LKERILQLDL ENKTLALAAS SRSPLDSHGE ESSLDVNVLK DKMEKLKRLL QVAARKSQVT LDVEKLCDLE IMPSSEAADG EKATALYYQQ ELKQLKEEFE RYKMRAQVVL KSKNTKDGNL GKELEAAQEQ LAELKEKYIS LRLSCEELEH QHQQEADDWK QELARLQQLH RQELERCQLD FRDRTLKLEE ELHKQRDRAL AVLTEKDLEL EQLRSVALAS GLPGRRSPVG GGGPGDPADT SSSDSLTQAL QLAAANEPTF FLYAEQLARK EVEITSLRKQ KHRLEVEVHQ LQDRLLEEGE RHREEVAALQ SHIEKNIRDQ SREGANLEYL KNIIYRFLTL PDSLGRQQTL TAILTILHFS PEEKQVIMRL PTSASWWPSG KR | |
Greater than 90% as determined by SDS-PAGE. |
79571 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |